Product Details

SNP ID
rs138995196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69632897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTCCCCCTTCAATGACCTCAA[C/T]CGGCAGCTGGTGAACATGGGCTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf35 PubMed Links

Gene Details

Gene
C10orf35
Gene Name
chromosome 10 open reading frame 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145306.2 915 Silent Mutation AAC,AAT N68N NP_660349.1
XM_005269606.1 915 Silent Mutation AAC,AAT N110N XP_005269663.1
XM_005269608.3 915 Silent Mutation AAC,AAT N68N XP_005269665.1
XM_011539455.2 915 Silent Mutation AAC,AAT N68N XP_011537757.1
XM_017015836.1 915 Silent Mutation AAC,AAT N68N XP_016871325.1

View Full Product Details