Product Details

SNP ID
rs139228214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91908409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCGGTGCGCTTTGGGGAGGCGG[G/T]GTCCCAGCGGCTGGGCCGGACGCAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGFBP3 PubMed Links

Gene Details

Gene
FGFBP3
Gene Name
fibroblast growth factor binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152429.4 743 Silent Mutation ACA,ACC T187T NP_689642.3

View Full Product Details