Product Details

SNP ID
rs139938475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15213284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTTTCTGCTGGTGCTCTTGGAC[A/G]GGCGGGTAGTTCTGCTGCAGAGACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM171A1 PubMed Links

Gene Details

Gene
FAM171A1
Gene Name
family with sequence similarity 171 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010924.1 2232 Silent Mutation NP_001010924.1
XM_011519378.2 2232 Silent Mutation XP_011517680.1
XM_017015904.1 2232 Silent Mutation XP_016871393.1
XM_017015905.1 2232 Silent Mutation XP_016871394.1

View Full Product Details