Product Details

SNP ID
rs140388364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102854403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTAATCGCGCTCACCAAGCAGGT[C/G]CTGAAAGGCTCCCGGAGCTCCGAGG
Phenotype
MIM: 616600
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BORCS7 PubMed Links

Gene Details

Gene
BORCS7
Gene Name
BLOC-1 related complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136200.1 194 Silent Mutation GTC,GTG V39V NP_001129672.1
NM_144591.3 194 Silent Mutation GTC,GTG V39V NP_653192.2
Gene
BORCS7-ASMT
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details