Product Details

SNP ID
rs141168124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117137938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGGCTCCTTGAGGAAGGCGGCT[C/G]GGAGGTTCCCCTCCGCGCCCTTGCT
Phenotype
MIM: 604294
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VAX1 PubMed Links

Gene Details

Gene
VAX1
Gene Name
ventral anterior homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112704.1 364 Missense Mutation CCA,CGA P40R NP_001106175.1
NM_199131.2 364 Missense Mutation CCA,CGA P40R NP_954582.1

View Full Product Details