Product Details
- SNP ID
-
rs141506679
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:102470674 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCTTTCCCCGGCAGGTCAGGCGC[C/T]GGGCTCTCCTCAAGGGCTGTGGTGC
- Phenotype
-
MIM: 605143
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACTR1A
PubMed Links
Gene Details
- Gene
- ACTR1A
- Gene Name
- ARP1 actin-related protein 1 homolog A, centractin alpha
There are no transcripts associated with this gene.
- Gene
- MFSD13A
- Gene Name
- major facilitator superfamily domain containing 13A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024789.3 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
NP_079065.2 |
XM_006717973.3 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_006718036.1 |
XM_006717974.3 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_006718037.1 |
XM_011540160.2 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_011538462.1 |
XM_011540163.2 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_011538465.1 |
XM_011540167.2 |
547 |
Intron |
|
|
XP_011538469.1 |
XM_011540169.2 |
547 |
Intron |
|
|
XP_011538471.1 |
XM_017016654.1 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_016872143.1 |
XM_017016655.1 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_016872144.1 |
XM_017016656.1 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_016872145.1 |
XM_017016657.1 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_016872146.1 |
XM_017016658.1 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_016872147.1 |
XM_017016659.1 |
547 |
Intron |
|
|
XP_016872148.1 |
XM_017016660.1 |
547 |
Intron |
|
|
XP_016872149.1 |
XM_017016661.1 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_016872150.1 |
XM_017016662.1 |
547 |
Silent Mutation |
GCC,GCT |
A87A |
XP_016872151.1 |
View Full Product Details