Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001198798.2 | 801 | Missense Mutation | TAC,TGC | Y325C | NP_001185727.1 |
NM_001198799.2 | 801 | UTR 3 | NP_001185728.1 | ||
NM_001198800.2 | 801 | Missense Mutation | TAC,TGC | Y325C | NP_001185729.1 |
XM_006717873.3 | 801 | Missense Mutation | TAC,TGC | Y325C | XP_006717936.1 |
XM_011539841.2 | 801 | UTR 3 | XP_011538143.1 | ||
XM_011539842.2 | 801 | UTR 3 | XP_011538144.1 | ||
XM_017016291.1 | 801 | UTR 3 | XP_016871780.1 | ||
XM_017016292.1 | 801 | UTR 3 | XP_016871781.1 | ||
XM_017016293.1 | 801 | UTR 3 | XP_016871782.1 | ||
XM_017016294.1 | 801 | UTR 3 | XP_016871783.1 | ||
XM_017016295.1 | 801 | UTR 3 | XP_016871784.1 | ||
XM_017016296.1 | 801 | UTR 3 | XP_016871785.1 | ||
XM_017016297.1 | 801 | UTR 3 | XP_016871786.1 | ||
XM_017016298.1 | 801 | UTR 3 | XP_016871787.1 | ||
XM_017016299.1 | 801 | Missense Mutation | TAC,TGC | Y285C | XP_016871788.1 |
XM_017016300.1 | 801 | Missense Mutation | TAC,TGC | Y285C | XP_016871789.1 |
XM_017016301.1 | 801 | UTR 3 | XP_016871790.1 | ||
XM_017016302.1 | 801 | UTR 3 | XP_016871791.1 | ||
XM_017016303.1 | 801 | UTR 3 | XP_016871792.1 |