Product Details
- SNP ID
-
rs142221261
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:27158656 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCATTCATTGTCCATTTGTATTA[C/T]TCACTGCAGTCTGCAAACAATGTCT
- Phenotype
-
MIM: 608221
MIM: 607472
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MASTL
PubMed Links
Gene Details
- Gene
- MASTL
- Gene Name
- microtubule associated serine/threonine kinase like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001172303.2 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
NP_001165774.1 |
NM_001172304.2 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
NP_001165775.1 |
NM_001320756.1 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
NP_001307685.1 |
NM_001320757.1 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
NP_001307686.1 |
NM_032844.4 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
NP_116233.2 |
XM_005252631.3 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
XP_005252688.1 |
XM_005252632.3 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
XP_005252689.1 |
XM_006717519.3 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
XP_006717582.1 |
XM_006717520.3 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
XP_006717583.1 |
XM_011519751.2 |
1250 |
Intron |
|
|
XP_011518053.1 |
XM_017016852.1 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
XP_016872341.1 |
XM_017016853.1 |
1250 |
Silent Mutation |
TAC,TAT |
Y49Y |
XP_016872342.1 |
XM_017016854.1 |
1250 |
Intron |
|
|
XP_016872343.1 |
XM_017016855.1 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
XP_016872344.1 |
XM_017016856.1 |
1250 |
Silent Mutation |
TAC,TAT |
Y98Y |
XP_016872345.1 |
- Gene
- YME1L1
- Gene Name
- YME1 like 1 ATPase
There are no transcripts associated with this gene.
View Full Product Details