Product Details

SNP ID
rs143055894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79349745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCACAGGGTGATCCCTTCCTTC[A/G]TGTGCCAGGTAATGTAGTTTCCTCT
Phenotype
MIM: 604486
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPIF PubMed Links

Gene Details

Gene
PPIF
Gene Name
peptidylprolyl isomerase F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005729.3 390 Missense Mutation ATG,GTG M103V NP_005720.1
XM_005269379.2 390 Missense Mutation ATG,GTG M103V XP_005269436.1

View Full Product Details