Product Details

SNP ID
rs143390578
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93613148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTGTGTTTCCATTGGACATTGG[A/G]ATAGTGGGTTGGGCTGCTCACACGA
Phenotype
MIM: 600827
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PDE6C PubMed Links

Gene Details

Gene
PDE6C
Gene Name
phosphodiesterase 6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006204.3 561 Silent Mutation GGA,GGG G141G NP_006195.3
XM_017016322.1 561 Silent Mutation GGA,GGG G44G XP_016871811.1

View Full Product Details