Product Details

SNP ID
rs143796774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73751882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAGACTACCCTAAGGGAGAAAT[A/G]GGAAGAGGAAACTTCAGGATTATTT
Phenotype
MIM: 607185
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC24C PubMed Links

Gene Details

Gene
SEC24C
Gene Name
SEC24 homolog C, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004922.3 Intron NP_004913.2
NM_198597.2 Intron NP_940999.1
XM_011540380.2 Intron XP_011538682.1
XM_011540381.2 Intron XP_011538683.1
XM_017016967.1 Intron XP_016872456.1
XM_017016968.1 Intron XP_016872457.1

View Full Product Details