Product Details

SNP ID
rs144612408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43462311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTACGTGGCTTTCTAGCCACGTA[A/G]TCATTGATTACAGGCACACCTCGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF487 PubMed Links

Gene Details

Gene
ZNF487
Gene Name
zinc finger protein 487
There are no transcripts associated with this gene.

View Full Product Details