Product Details

SNP ID
rs145222143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119038294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTCTCTCTCTGTCCCTTTCTCT[C/T]TCAGGGTCCTTGTCATTCTCCTCCC
Phenotype
MIM: 602039 MIM: 608226
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3A PubMed Links

Gene Details

Gene
EIF3A
Gene Name
eukaryotic translation initiation factor 3 subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003750.2 3818 Silent Mutation GAA,GAG E1224E NP_003741.1
Gene
NANOS1
Gene Name
nanos C2HC-type zinc finger 1
There are no transcripts associated with this gene.

View Full Product Details