Product Details

SNP ID
rs145591492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93613191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACGAAGAAAACTCATAATGTC[C/T]CAGATGTGAAAAAGGTAGGTGGCCT
Phenotype
MIM: 600827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE6C PubMed Links

Gene Details

Gene
PDE6C
Gene Name
phosphodiesterase 6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006204.3 604 Missense Mutation CCA,TCA P156S NP_006195.3
XM_017016322.1 604 Missense Mutation CCA,TCA P59S XP_016871811.1

View Full Product Details