Product Details

SNP ID
rs145772897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14825563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCATATGCACACTCATTGGGC[A/G]AGTGACTTCACTTAGGATCTTTGTG
Phenotype
MIM: 611233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDNF PubMed Links

Gene Details

Gene
CDNF
Gene Name
cerebral dopamine neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029954.2 339 Missense Mutation CGC,TGC R101C NP_001025125.2
XM_011519488.2 339 Missense Mutation CGC,TGC R101C XP_011517790.1

View Full Product Details