Product Details

SNP ID
rs145922405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:67925573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAATTTATCTCTCCTTTTAAAA[A/G]TATGGTACTAAGCAACACTAAAAAT
Phenotype
MIM: 609248 MIM: 604479
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HERC4 PubMed Links

Gene Details

Gene
HERC4
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278185.1 Intron NP_001265114.1
NM_001278186.1 Intron NP_001265115.1
NM_001278187.1 Intron NP_001265116.1
NM_015601.3 Intron NP_056416.2
NM_022079.2 Intron NP_071362.1
XM_011539592.2 Intron XP_011537894.1
XM_011539593.2 Intron XP_011537895.1
XM_011539594.2 Intron XP_011537896.1
XM_011539595.2 Intron XP_011537897.1
XM_011539596.2 Intron XP_011537898.1
XM_011539597.2 Intron XP_011537899.1
XM_017016041.1 Intron XP_016871530.1
Gene
SIRT1
Gene Name
sirtuin 1
There are no transcripts associated with this gene.

View Full Product Details