Product Details
- SNP ID
-
rs145940605
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:69404904 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCTGATCCATCCTGGGGACGCCC[C/T]GCCTCCCCACTGGTAGCCTCGGAGG
- Phenotype
-
MIM: 142600
MIM: 162321
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2229170] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HK1
- Gene Name
- hexokinase 1
There are no transcripts associated with this gene.
- Gene
- TACR2
- Gene Name
- tachykinin receptor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001057.2 |
1714 |
Silent Mutation |
GCA,GCG |
A373A |
NP_001048.2 |
View Full Product Details