Product Details

SNP ID
rs145979821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72274351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACCCGGGAGGAGGGGTTTGACCG[C/T]TCCACGAGCCTGGAGAGCTCGGACT
Phenotype
MIM: 607729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDIT4 PubMed Links

Gene Details

Gene
DDIT4
Gene Name
DNA damage inducible transcript 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019058.2 337 Silent Mutation CGC,CGT R45R NP_061931.1

View Full Product Details