Product Details
- SNP ID
-
rs146887700
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:67923092 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGTGGACTGGATGACTAGTTTCA[C/G]ACTCTTCATACCAAGAATAGGAATG
- Phenotype
-
MIM: 609248
MIM: 604479
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HERC4
PubMed Links
Gene Details
- Gene
- HERC4
- Gene Name
- HECT and RLD domain containing E3 ubiquitin protein ligase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278185.1 |
3374 |
Missense Mutation |
CTG,GTG |
L927V |
NP_001265114.1 |
NM_001278186.1 |
3374 |
Missense Mutation |
CTG,GTG |
L742V |
NP_001265115.1 |
NM_001278187.1 |
3374 |
Intron |
|
|
NP_001265116.1 |
NM_015601.3 |
3374 |
Missense Mutation |
CTG,GTG |
L997V |
NP_056416.2 |
NM_022079.2 |
3374 |
Missense Mutation |
CTG,GTG |
L1005V |
NP_071362.1 |
XM_011539592.2 |
3374 |
Missense Mutation |
CTG,GTG |
L1029V |
XP_011537894.1 |
XM_011539593.2 |
3374 |
Missense Mutation |
CTG,GTG |
L1021V |
XP_011537895.1 |
XM_011539594.2 |
3374 |
Missense Mutation |
CTG,GTG |
L959V |
XP_011537896.1 |
XM_011539595.2 |
3374 |
Missense Mutation |
CTG,GTG |
L959V |
XP_011537897.1 |
XM_011539596.2 |
3374 |
Missense Mutation |
CTG,GTG |
L935V |
XP_011537898.1 |
XM_011539597.2 |
3374 |
Missense Mutation |
CTG,GTG |
L908V |
XP_011537899.1 |
XM_017016041.1 |
3374 |
Missense Mutation |
CTG,GTG |
L884V |
XP_016871530.1 |
- Gene
- SIRT1
- Gene Name
- sirtuin 1
There are no transcripts associated with this gene.
View Full Product Details