Product Details

SNP ID
rs146974174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69404300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGGAGTTCTGGCCCCATGTT[A/G]TCTTTGTATTGGTATACTGAAGGAT
Phenotype
MIM: 142600 MIM: 162321
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HK1 PubMed Links

Gene Details

Gene
HK1
Gene Name
hexokinase 1
There are no transcripts associated with this gene.

Gene
TACR2
Gene Name
tachykinin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001057.2 2318 UTR 3 NP_001048.2

View Full Product Details