Product Details

SNP ID
rs147705683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133326886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTGGCCCCCTTACCCGCAGCA[A/G]GAAGCCGTCGGGGCAGGTGAACTGG
Phenotype
MIM: 604647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALY PubMed Links

Gene Details

Gene
CALY
Gene Name
calcyon neuron specific vesicular protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321617.1 1654 UTR 5 NP_001308546.1
NM_015722.3 1654 Silent Mutation CTG,TTG L118L NP_056537.1
XM_005252685.4 1654 Silent Mutation CTG,TTG L151L XP_005252742.2
XM_011539836.2 1654 Silent Mutation CTG,TTG L151L XP_011538138.1
XM_011539838.2 1654 Silent Mutation CTG,TTG L31L XP_011538140.1

View Full Product Details