Product Details

SNP ID
rs147741849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73803272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGACAGCGGTTCTGCAGGGAGC[A/G]TAGTGCCAGAGGGGTCTGGGAGGAG
Phenotype
MIM: 602123 MIM: 603268
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMK2G PubMed Links

Gene Details

Gene
CAMK2G
Gene Name
calcium/calmodulin dependent protein kinase II gamma
There are no transcripts associated with this gene.

Gene
NDST2
Gene Name
N-deacetylase and N-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003635.3 3034 Missense Mutation CGC,TGC R744C NP_003626.1
XM_005270255.3 3034 Missense Mutation CGC,TGC R744C XP_005270312.1
XM_005270256.3 3034 Missense Mutation CGC,TGC R370C XP_005270313.1
XM_011540310.2 3034 Missense Mutation CGC,TGC R744C XP_011538612.1
XM_017016857.1 3034 Missense Mutation CGC,TGC R267C XP_016872346.1
Gene
ZSWIM8
Gene Name
zinc finger SWIM-type containing 8
There are no transcripts associated with this gene.

Gene
ZSWIM8-AS1
Gene Name
ZSWIM8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details