Product Details

SNP ID
rs147899211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73136897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCATCATCCAGATCATCAGAA[G/T]CTGACTCATTAGGCCTTGGCCCTAC
Phenotype
MIM: 616464 MIM: 609233
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ECD PubMed Links

Gene Details

Gene
ECD
Gene Name
ecdysoneless cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135752.1 1853 Missense Mutation GAT,GCT D537A NP_001129224.1
NM_001135753.1 1853 Missense Mutation GAT,GCT D461A NP_001129225.1
NM_007265.2 1853 Missense Mutation GAT,GCT D504A NP_009196.1
Gene
NUDT13
Gene Name
nudix hydrolase 13
There are no transcripts associated with this gene.

View Full Product Details