Product Details

SNP ID
rs148066730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73012726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAACAAAAAGATGTAATTCAAA[C/T]AGGGCTAAGGAATGGAAAAAGTGAT
Phenotype
MIM: 176710
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P4HA1 PubMed Links

Gene Details

Gene
P4HA1
Gene Name
prolyl 4-hydroxylase subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000917.3 Intron NP_000908.2
NM_001017962.2 Intron NP_001017962.1
NM_001142595.1 Intron NP_001136067.1
NM_001142596.1 Intron NP_001136068.1

View Full Product Details