Product Details

SNP ID
rs148469925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125721374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAACTAGGGCCAGCAGGAACTAC[C/G]ATTCTTGGCAACAGCAAGAAAAGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EDRF1 PubMed Links

Gene Details

Gene
EDRF1
Gene Name
erythroid differentiation regulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202438.1 572 Silent Mutation ACC,ACG T93T NP_001189367.1
NM_015608.2 572 Silent Mutation ACC,ACG T93T NP_056423.2
XM_011539622.1 572 Intron XP_011537924.1
XM_017016066.1 572 UTR 5 XP_016871555.1
Gene
FLJ37035
Gene Name
uncharacterized LOC399821
There are no transcripts associated with this gene.

View Full Product Details