Product Details

SNP ID
rs149325838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110498258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGTGCGCGGCTCGCTCAACGTCA[A/G]CCTCAACTCGGTGGTGCTGCGGCGG
Phenotype
MIM: 603069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP5 PubMed Links

Gene Details

Gene
DUSP5
Gene Name
dual specificity phosphatase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004419.3 392 Missense Mutation AAC,AGC N46S NP_004410.3

View Full Product Details