Product Details

SNP ID
rs149350791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73802482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACTGGAATGCTGCAGTTCTTCC[C/T]GAAGCCACGAGGGCACTGGCTGTCC
Phenotype
MIM: 603268
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDST2 PubMed Links

Gene Details

Gene
NDST2
Gene Name
N-deacetylase and N-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003635.3 3388 Missense Mutation CAG,CGG Q874R NP_003626.1
XM_005270255.3 3388 UTR 3 XP_005270312.1
XM_005270256.3 3388 Missense Mutation CAG,CGG Q500R XP_005270313.1
XM_011540310.2 3388 Missense Mutation CAG,CGG Q874R XP_011538612.1
XM_017016857.1 3388 Missense Mutation CAG,CGG Q397R XP_016872346.1
Gene
ZSWIM8
Gene Name
zinc finger SWIM-type containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242487.1 3388 Intron NP_001229416.1
NM_001242488.1 3388 Intron NP_001229417.1
NM_015037.3 3388 Intron NP_055852.2
XM_005269648.2 3388 Intron XP_005269705.1
XM_005269653.3 3388 Intron XP_005269710.1
XM_005269655.3 3388 Intron XP_005269712.1
XM_006717725.3 3388 Intron XP_006717788.1
XM_011539544.1 3388 Intron XP_011537846.1
XM_017015978.1 3388 Intron XP_016871467.1
Gene
ZSWIM8-AS1
Gene Name
ZSWIM8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details