Product Details

SNP ID
rs150206121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124464455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGAAGCCGGAGGTGTTTGGGAG[A/G]CTGGGAACTGGGAACAGGGAGACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LHPP PubMed Links

Gene Details

Gene
LHPP
Gene Name
phospholysine phosphohistidine inorganic pyrophosphate phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167880.1 Intron NP_001161352.1
NM_001318331.1 Intron NP_001305260.1
NM_001318332.1 Intron NP_001305261.1
NM_022126.3 Intron NP_071409.3
XM_005270026.3 Intron XP_005270083.1
XM_006717943.1 Intron XP_006718006.1
XM_011540058.2 Intron XP_011538360.1
XM_017016509.1 Intron XP_016871998.1
XM_017016510.1 Intron XP_016871999.1
XM_017016511.1 Intron XP_016872000.1
XM_017016512.1 Intron XP_016872001.1

View Full Product Details