Product Details
- SNP ID
-
rs2512371
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:87044280 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGGCTTGAAAACCACTGATTGAA[G/A]AAATCAGTTCAGTGATACAGACTGA
- Phenotype
-
MIM: 616360
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM135
PubMed Links
Gene Details
- Gene
- TMEM135
- Gene Name
- transmembrane protein 135
There are no transcripts associated with this gene.
View Full Product Details