Product Details
- SNP ID
-
rs111564700
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:93332285 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGAGAACCAAGCCCATAATACGA[C/T]GGGGTAAGTGCTGAGAAATTTCTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEUP1
PubMed Links
Gene Details
- Gene
- DEUP1
- Gene Name
- deuterosome assembly protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181645.3 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
NP_857596.2 |
XM_005273802.1 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_005273859.1 |
XM_011542629.2 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_011540931.1 |
XM_011542630.1 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_011540932.1 |
XM_011542631.2 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_011540933.1 |
XM_011542632.2 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_011540934.1 |
XM_011542633.2 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_011540935.1 |
XM_011542635.2 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_011540937.1 |
XM_011542636.2 |
173 |
Intron |
|
|
XP_011540938.1 |
XM_011542637.2 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_011540939.1 |
XM_011542638.2 |
173 |
Intron |
|
|
XP_011540940.1 |
XM_017017245.1 |
173 |
Missense Mutation |
ACG,ATG |
T9M |
XP_016872734.1 |
- Gene
- LOC102723746
- Gene Name
- uncharacterized LOC102723746
There are no transcripts associated with this gene.
View Full Product Details