Product Details

SNP ID
rs113305638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61816858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGATTTGCTGGCGCGCGCCCAG[A/C]GCCAGCCGCCTGCGCGCCGGGTTTT
Phenotype
MIM: 606148 MIM: 606149
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013402.4 200 Silent Mutation GCG,GCT A24A NP_037534.3
XM_011545022.2 200 Intron XP_011543324.1
Gene
FADS2
Gene Name
fatty acid desaturase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281501.1 200 Intron NP_001268430.1
NM_001281502.1 200 Intron NP_001268431.1
NM_004265.3 200 Intron NP_004256.1
Gene
MIR1908
Gene Name
microRNA 1908
There are no transcripts associated with this gene.

View Full Product Details