Product Details

SNP ID
rs113605041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64935492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTGGGGAGATGTCTCTACGTG[G/T]GTGTGCAGGTGGGATGGCGAGTTCT
Phenotype
MIM: 609651 MIM: 601644
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPHA2 PubMed Links

Gene Details

Gene
GPHA2
Gene Name
glycoprotein hormone alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130769.3 Intron NP_570125.1
XM_011544774.2 Intron XP_011543076.1
XM_011544775.2 Intron XP_011543077.1
XM_011544776.2 Intron XP_011543078.1
Gene
MAJIN
Gene Name
membrane anchored junction protein
There are no transcripts associated with this gene.

Gene
PPP2R5B
Gene Name
protein phosphatase 2 regulatory subunit B'beta
There are no transcripts associated with this gene.

View Full Product Details