Product Details
- SNP ID
-
rs114437963
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:81888185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATCTAGTAAATGCTGGGTTTAGG[G/T]TTTCTCCTGAAGTCTAACCTAACCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4300
PubMed Links
Gene Details
- Gene
- MIR4300
- Gene Name
- microRNA 4300
There are no transcripts associated with this gene.
- Gene
- MIR4300HG
- Gene Name
- MIR4300 host gene
There are no transcripts associated with this gene.
View Full Product Details