Product Details

SNP ID
rs114746256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74842659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAGTTTGCTCCAATTTACTGTC[A/G]TGTTCACATTCAACCATTCATGCTC
Phenotype
MIM: 609788
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF169 PubMed Links

Gene Details

Gene
RNF169
Gene Name
ring finger protein 169
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098638.1 2990 Intron NP_001092108.1
XM_011544889.2 2990 Intron XP_011543191.1
Gene
XRRA1
Gene Name
X-ray radiation resistance associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270380.1 2990 UTR 3 NP_001257309.1
NM_001270381.1 2990 UTR 3 NP_001257310.1
NM_182969.2 2990 UTR 3 NP_892014.1
XM_005273765.2 2990 UTR 3 XP_005273822.1
XM_005273770.2 2990 UTR 3 XP_005273827.1
XM_011544755.1 2990 UTR 3 XP_011543057.1
XM_011544756.1 2990 UTR 3 XP_011543058.1
XM_011544757.1 2990 UTR 3 XP_011543059.1
XM_011544758.1 2990 UTR 3 XP_011543060.1
XM_011544759.1 2990 UTR 3 XP_011543061.1
XM_011544760.2 2990 Intron XP_011543062.1
XM_011544761.1 2990 UTR 3 XP_011543063.1
XM_011544762.1 2990 UTR 3 XP_011543064.1
XM_011544763.1 2990 UTR 3 XP_011543065.1
XM_011544764.1 2990 UTR 3 XP_011543066.1
XM_011544765.1 2990 UTR 3 XP_011543067.1
XM_011544766.2 2990 UTR 3 XP_011543068.1
XM_011544767.1 2990 UTR 3 XP_011543069.1
XM_011544768.1 2990 UTR 3 XP_011543070.1
XM_017017221.1 2990 UTR 3 XP_016872710.1
XM_017017222.1 2990 UTR 3 XP_016872711.1
XM_017017223.1 2990 UTR 3 XP_016872712.1
XM_017017224.1 2990 UTR 3 XP_016872713.1
XM_017017225.1 2990 UTR 3 XP_016872714.1
XM_017017226.1 2990 UTR 3 XP_016872715.1
XM_017017227.1 2990 UTR 3 XP_016872716.1
XM_017017228.1 2990 UTR 3 XP_016872717.1
XM_017017229.1 2990 UTR 3 XP_016872718.1
XM_017017230.1 2990 UTR 3 XP_016872719.1
XM_017017231.1 2990 UTR 3 XP_016872720.1
XM_017017232.1 2990 UTR 3 XP_016872721.1
XM_017017233.1 2990 UTR 3 XP_016872722.1
XM_017017234.1 2990 UTR 3 XP_016872723.1

View Full Product Details