Product Details

SNP ID
rs114921537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62271035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGAATGTTTTCTTAACCAAAC[A/G]GATGAAACTCTGAGCAATGTTGAGG
Phenotype
MIM: 605562
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCGB2A2 PubMed Links

Gene Details

Gene
SCGB2A2
Gene Name
secretoglobin family 2A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002411.3 297 Silent Mutation ACA,ACG T70T NP_002402.1
XM_005274005.3 297 Silent Mutation ACA,ACG T70T XP_005274062.3

View Full Product Details