Product Details

SNP ID
rs116658745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74842704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTCCTAATGAAGGAACCAAGAA[A/G]GAGTCTGAGTAAAGAACGAATATAC
Phenotype
MIM: 609788
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF169 PubMed Links

Gene Details

Gene
RNF169
Gene Name
ring finger protein 169
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098638.1 2945 Intron NP_001092108.1
XM_011544889.2 2945 Intron XP_011543191.1
Gene
XRRA1
Gene Name
X-ray radiation resistance associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270380.1 2945 UTR 3 NP_001257309.1
NM_001270381.1 2945 UTR 3 NP_001257310.1
NM_182969.2 2945 UTR 3 NP_892014.1
XM_005273765.2 2945 UTR 3 XP_005273822.1
XM_005273770.2 2945 UTR 3 XP_005273827.1
XM_011544755.1 2945 UTR 3 XP_011543057.1
XM_011544756.1 2945 UTR 3 XP_011543058.1
XM_011544757.1 2945 UTR 3 XP_011543059.1
XM_011544758.1 2945 UTR 3 XP_011543060.1
XM_011544759.1 2945 UTR 3 XP_011543061.1
XM_011544760.2 2945 Intron XP_011543062.1
XM_011544761.1 2945 UTR 3 XP_011543063.1
XM_011544762.1 2945 UTR 3 XP_011543064.1
XM_011544763.1 2945 UTR 3 XP_011543065.1
XM_011544764.1 2945 UTR 3 XP_011543066.1
XM_011544765.1 2945 UTR 3 XP_011543067.1
XM_011544766.2 2945 UTR 3 XP_011543068.1
XM_011544767.1 2945 UTR 3 XP_011543069.1
XM_011544768.1 2945 UTR 3 XP_011543070.1
XM_017017221.1 2945 UTR 3 XP_016872710.1
XM_017017222.1 2945 UTR 3 XP_016872711.1
XM_017017223.1 2945 UTR 3 XP_016872712.1
XM_017017224.1 2945 UTR 3 XP_016872713.1
XM_017017225.1 2945 UTR 3 XP_016872714.1
XM_017017226.1 2945 UTR 3 XP_016872715.1
XM_017017227.1 2945 UTR 3 XP_016872716.1
XM_017017228.1 2945 UTR 3 XP_016872717.1
XM_017017229.1 2945 UTR 3 XP_016872718.1
XM_017017230.1 2945 UTR 3 XP_016872719.1
XM_017017231.1 2945 UTR 3 XP_016872720.1
XM_017017232.1 2945 UTR 3 XP_016872721.1
XM_017017233.1 2945 UTR 3 XP_016872722.1
XM_017017234.1 2945 UTR 3 XP_016872723.1

View Full Product Details