Product Details

SNP ID
rs116960344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55984379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATGCCTTGGAGTTCCTTAATT[A/C]ATATTTATCACCTCATGCTTTACTA
Phenotype
MIM: 608492
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5F1 PubMed Links

Gene Details

Gene
OR5F1
Gene Name
olfactory receptor family 5 subfamily F member 1
There are no transcripts associated with this gene.

Gene
OR7E5P
Gene Name
olfactory receptor family 7 subfamily E member 5 pseudogene
There are no transcripts associated with this gene.

View Full Product Details