Product Details

SNP ID
rs116985986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93335513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAGGAAGATCTTCACTATAATT[A/G]TAGTTTGTCTAGTCCTTCTTGTATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEUP1 PubMed Links

Gene Details

Gene
DEUP1
Gene Name
deuterosome assembly protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181645.3 Intron NP_857596.2
XM_005273802.1 Intron XP_005273859.1
XM_011542629.2 Intron XP_011540931.1
XM_011542630.1 Intron XP_011540932.1
XM_011542631.2 Intron XP_011540933.1
XM_011542632.2 Intron XP_011540934.1
XM_011542633.2 Intron XP_011540935.1
XM_011542635.2 Intron XP_011540937.1
XM_011542636.2 Intron XP_011540938.1
XM_011542637.2 Intron XP_011540939.1
XM_011542638.2 Intron XP_011540940.1
XM_017017245.1 Intron XP_016872734.1
Gene
LOC102723746
Gene Name
uncharacterized LOC102723746
There are no transcripts associated with this gene.

View Full Product Details