Product Details

SNP ID
rs138558402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59109664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTGAAGAAAACAAGTCATTTAG[C/T]GCTATGGAAGATGACCAGAGGACTA
Phenotype
MIM: 615584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM111B PubMed Links

Gene Details

Gene
FAM111B
Gene Name
family with sequence similarity 111 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142703.1 282 Intron NP_001136175.1
NM_001142704.1 282 Intron NP_001136176.1
NM_198947.3 282 Silent Mutation AGC,AGT S13S NP_945185.1

View Full Product Details