Product Details

SNP ID
rs138681657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124424129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGTCCCTGACCCAAAAAACAGA[A/G]CAACAGCAATTATGTGGGAGCCCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8B4 PubMed Links

Gene Details

Gene
OR8B4
Gene Name
olfactory receptor family 8 subfamily B member 4 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005196.1 743 Missense Mutation GCT,GTT A248V NP_001005196.1

View Full Product Details