Product Details

SNP ID
rs138778962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118080821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTTCAGGCCTGAAAATCTCCA[C/T]GGCCTCAGACTTCCCAGGTGCCTCT
Phenotype
MIM: 606565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS4 PubMed Links

Gene Details

Gene
TMPRSS4
Gene Name
transmembrane protease, serine 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083947.1 Intron NP_001077416.1
NM_001173551.1 Intron NP_001167022.1
NM_001173552.1 Intron NP_001167023.1
NM_001290094.1 Intron NP_001277023.1
NM_001290096.1 Intron NP_001277025.1
NM_019894.3 Intron NP_063947.1
XM_005271613.3 Intron XP_005271670.1
XM_005271614.2 Intron XP_005271671.1
XM_005271615.3 Intron XP_005271672.1
XM_011542901.2 Intron XP_011541203.1
XM_011542902.2 Intron XP_011541204.1
XM_011542903.2 Intron XP_011541205.1
XM_011542904.2 Intron XP_011541206.1
Gene
TMPRSS4-AS1
Gene Name
TMPRSS4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details