Product Details

SNP ID
rs138821110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73308982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCCGCGGCCGCGGAAGAAGCGG[C/T]CGAGGGCCCAGCGCGAGGAGCCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF17 PubMed Links

Gene Details

Gene
ARHGEF17
Gene Name
Rho guanine nucleotide exchange factor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014786.3 365 Missense Mutation GCC,GTC A115V NP_055601.2
XM_017018623.1 365 Intron XP_016874112.1
XM_017018624.1 365 Intron XP_016874113.1

View Full Product Details