Product Details

SNP ID
rs138942341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113909313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCAGGAATTCTAGCCACAGATA[A/C]ACATCATCCCCAGGATTCTGCTCTG
Phenotype
MIM: 604654
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HTR3B PubMed Links

Gene Details

Gene
HTR3B
Gene Name
5-hydroxytryptamine receptor 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006028.4 322 Missense Mutation AAA,ACA K24T NP_006019.1
XM_011543063.1 322 Missense Mutation AAA,ACA K13T XP_011541365.1
XM_017018552.1 322 Intron XP_016874041.1

View Full Product Details