Product Details

SNP ID
rs139153091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55935935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGATGTGTGAACCAGGGAACTCA[C/T]GTTGCCTCCAAGGTATGACAAGACA
Phenotype
MIM: 608496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5I1 PubMed Links

Gene Details

Gene
OR5I1
Gene Name
olfactory receptor family 5 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006637.1 466 Missense Mutation ATG,GTG M156V NP_006628.1

View Full Product Details