Product Details

SNP ID
rs139264769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2159958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTCCAGGGCCAAGGGCTGCAGG[C/T]TGCCTGCACCAGGGCCCCCGCCCAG
Phenotype
MIM: 176730 MIM: 191290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INS PubMed Links

Gene Details

Gene
INS
Gene Name
insulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000207.2 312 Missense Mutation AAC,AGC N76S NP_000198.1
NM_001185097.1 312 Missense Mutation AAC,AGC N76S NP_001172026.1
NM_001185098.1 312 Missense Mutation AAC,AGC N76S NP_001172027.1
NM_001291897.1 312 Missense Mutation AAC,AGC N76S NP_001278826.1
Gene
INS-IGF2
Gene Name
INS-IGF2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042376.2 312 Intron NP_001035835.1
Gene
TH
Gene Name
tyrosine hydroxylase
There are no transcripts associated with this gene.

View Full Product Details