Product Details
- SNP ID
-
rs139595894
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59856030 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTTGGCAATTCCAGTCATTTTCA[C/T]TATAATAGTCTGATGATACAAAGAG
- Phenotype
-
MIM: 189905
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TCN1
PubMed Links
Gene Details
- Gene
- TCN1
- Gene Name
- transcobalamin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001062.3 |
874 |
Missense Mutation |
AAT,AGT |
N259S |
NP_001053.2 |
View Full Product Details