Product Details
- SNP ID
-
rs139627540
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:128462397 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCTGGCTTGACGTCCAGCATGGC[A/G]TGCAGCTCCTCAGGGGTGTACCCCA
- Phenotype
-
MIM: 164720
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ETS1
PubMed Links
Gene Details
- Gene
- ETS1
- Gene Name
- ETS proto-oncogene 1, transcription factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143820.1 |
1281 |
Silent Mutation |
CAC,CAT |
H474H |
NP_001137292.1 |
NM_001162422.1 |
1281 |
Silent Mutation |
CAC,CAT |
H214H |
NP_001155894.1 |
NM_005238.3 |
1281 |
Silent Mutation |
CAC,CAT |
H430H |
NP_005229.1 |
XM_011542649.2 |
1281 |
Silent Mutation |
CAC,CAT |
H508H |
XP_011540951.1 |
XM_011542650.1 |
1281 |
Silent Mutation |
CAC,CAT |
H464H |
XP_011540952.1 |
XM_011542651.1 |
1281 |
Silent Mutation |
CAC,CAT |
H421H |
XP_011540953.1 |
XM_011542652.1 |
1281 |
Silent Mutation |
CAC,CAT |
H343H |
XP_011540954.1 |
XM_017017314.1 |
1281 |
Silent Mutation |
CAC,CAT |
H474H |
XP_016872803.1 |
XM_017017315.1 |
1281 |
Silent Mutation |
CAC,CAT |
H439H |
XP_016872804.1 |
XM_017017316.1 |
1281 |
Silent Mutation |
CAC,CAT |
H407H |
XP_016872805.1 |
XM_017017317.1 |
1281 |
Silent Mutation |
CAC,CAT |
H387H |
XP_016872806.1 |
View Full Product Details