Product Details

SNP ID
rs139636004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57712722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGTCGGCCCTGCTCTCTGCTG[C/T]CTTCCTACTCGTGAGGAAACTGCCG
Phenotype
MIM: 612385 MIM: 616715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED19 PubMed Links

Gene Details

Gene
MED19
Gene Name
mediator complex subunit 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317078.1 200 Intron NP_001304007.1
NM_153450.2 200 Intron NP_703151.2
Gene
TMX2
Gene Name
thioredoxin related transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144012.2 200 Missense Mutation GCC,GTC A35V NP_001137484.1
NM_015959.3 200 Missense Mutation GCC,GTC A35V NP_057043.1
Gene
TMX2-CTNND1
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details