Product Details

SNP ID
rs139901573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56290474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAACCAGCTAAAATGTGTATCA[A/T]GATTTCAATGTCGTATGTGTCCATG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8H1 PubMed Links

Gene Details

Gene
OR8H1
Gene Name
olfactory receptor family 8 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005199.1 Intron NP_001005199.1

View Full Product Details