Product Details

SNP ID
rs140345028
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123977003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCCACGATGAAGATGTAGGAA[A/G]TAACGATGAGGATGAGGCAGCCTGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR10S1 PubMed Links

Gene Details

Gene
OR10S1
Gene Name
olfactory receptor family 10 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004474.1 689 Missense Mutation ACT,ATT T230I NP_001004474.1

View Full Product Details